Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

772879

Sigma-Aldrich

Yttrium aluminum borate

powder, -200 mesh

Synonym(s):

Aluminum yttrium borate, YAB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
YAl3(BO3)4
CAS Number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

assay

99.9% trace metals basis

form

powder

particle size

-200 mesh

mp

>400 °C

SMILES string

[Al+3].[Al+3].[Al+3].[Y+3].[O-]B([O-])[O-].[O-]B([O-])[O-].[O-]B([O-])[O-].[O-]B([O-])[O-]

InChI

1S/3Al.4BO3.Y/c;;;4*2-1(3)4;/q3*+3;4*-3;+3

InChI key

XKVPKGQXTWIKDR-UHFFFAOYSA-N

Application

YAB is a promising phosphor host material, showing different emission properties upon doping with rare earth and other elements. Doped YAB crystals are widely studied for their non-linear optical properties and are used for laser and UV applications.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiang Liu et al.
Optics letters, 36(14), 2653-2655 (2011-07-19)
A yttrium aluminum borate [YAl(3)(BO(3))(4)] (YAB) crystal with UV cutoff wavelength of 165 nm is used as the nonlinear optical crystal for fourth harmonic generation. The fundamental frequency laser at 1064 nm from an Nd:YVO(4) master oscillator power amplifier laser
Jaque, D.;
Journal of Applied Physics, 75(3), 325-325 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service