Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

772747

Sigma-Aldrich

6-Chloro-2-methoxypyridine-3-carboxaldehyde

97%

Synonym(s):

6-Chloro-2-methoxy-3-pyridinecarbaldehyde, 6-Chloro-2-methoxynicotinaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6ClNO2
CAS Number:
Molecular Weight:
171.58
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

78-81 °C

functional group

aldehyde
chloro

SMILES string

COc1nc(Cl)ccc1C=O

InChI

1S/C7H6ClNO2/c1-11-7-5(4-10)2-3-6(8)9-7/h2-4H,1H3

InChI key

AVBARORPQMEWPR-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service