Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

772666

Sigma-Aldrich

Lanthanum calcium borate

powder, -200 mesh, 99.9% trace metals basis

Synonym(s):

Boron calcium lanthanum oxide, Calcium Lanthanum Borate, LCBO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
La2CaB10O19
CAS Number:
UNSPSC Code:
26111700
NACRES:
NA.23

assay

99.9% trace metals basis

form

powder

particle size

-200 mesh

mp

>300 °C

Application

LCBO is a promising phosphor host material, showing different emission properties upon doping with rare earth and other elements. LCBO crystals are also studied for piezoelectric properties.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brenier, A.;
Journal of Applied Physics, 108(9), 093101/1-093101/1 (2010)
Ozga, K.;
Journal of Crystal Growth, 344(1), 27-27 (2012)

Articles

A significant limiting factor for wearable electronics and wireless sensors is the finite amount of energy that can be stored in on-board batteries.

Among various ceramics, one-dimensional (1-D) piezoelectric ceramics have attracted significant scientific attention for use in energy harvesting.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service