Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

772658

Sigma-Aldrich

Barium pyrophosphate

powder, -200 mesh, 99.9% trace metals basis

Synonym(s):

Barium diphosphate, Barium phosphate, Dibarium diphosphate, Dibarium pyrophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ba2P2O7
CAS Number:
Molecular Weight:
448.60
EC Number:
UNSPSC Code:
26111700

assay

99.9% trace metals basis

form

powder

particle size

-200 mesh

InChI

1S/2Ba.H4O7P2/c;;1-8(2,3)7-9(4,5)6/h;;(H2,1,2,3)(H2,4,5,6)/q2*+2;/p-4

InChI key

RCUAPGYXYWSYKO-UHFFFAOYSA-J

Application

Ba2P2O7 is a promising phosphor host material, showing different emission properties upon doping with rare earth and other elements. Bi doped Ba2P2O7 crystals exhibit broadband NIR photoluminescence.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mingying Peng et al.
Optics express, 18(12), 12852-12863 (2010-07-01)
We report on a novel type of Bi-doped crystal that exhibits ultrabroadband photoluminescence in the near infrared (NIR). Emission centers can be generated and degenerated reversibly by annealing the material in CO atmosphere and air, respectively, indicating that emission is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service