Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

772542

Sigma-Aldrich

Poly(hydroxyethyl methacrylate), DDMAT terminated

average Mn 7,000, PDI <1.2

Synonym(s):

Macro RAFT-CTA, PHEA, PHEMA, PolyHEA, PolyHEMA, RAFT polymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C4H7O2(C6H10O3)nC13H25S3
UNSPSC Code:
12352100
NACRES:
NA.23

form

solid

mol wt

average Mn 7,000

impurities

≤10% Diethyl ether

transition temp

Tm 167-172 °C

PDI

<1.2

Application

Poly(hydroxyethyl methacrylate) is a hydrophobic polymer which contains a hydrophilic hydroxy group. The end group functionality contains a carboxylic acid (COOH) as well as the 2-(Dodecylthiocarbonothioylthio)-2-methylpropionic acid RAFT agent moiety. This will allow for additional polymerization to generate a diblock copolymer. This RAFT agent is especially suited for the polymerization of styrene; acrylate and acrylamide monomers.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Reversible addition–fragmentation chain transfer (RAFT) polymerization is rapidly moving to the forefront in construction of drug and gene delivery vehicles.

Professor Shrike Zhang (Harvard Medical School, USA) discusses advances in 3D-bioprinted tissue models for in vitro drug testing, reviews bioink selections, and provides application examples of 3D bioprinting in tissue model biofabrication.

Over the past two decades, the rapid advance of controlled living polymerization (CLP) techniques.

Microparticles in drug delivery: Study on controlling chitosan microparticle size and distribution, exploring encapsulation of BSA and TPP cross-linker.

See All

Protocols

Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service