Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

772402

Sigma-Aldrich

PBDTTT-CF

Synonym(s):

Poly[1-(6-{4,8-bis[(2-ethylhexyl)oxy]-6-methylbenzo[1,2-b:4,5-b′]dithiophen-2-yl}-3-fluoro-4-methylthieno[3,4-b]thiophen-2-yl)-1-octanone]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H51FO3S4 ]n
CAS Number:
Molecular Weight:
727.09 (as monomer)
UNSPSC Code:
12352103
NACRES:
NA.23

description

Band gap: 1.77 eV

form

powder

mol wt

average Mw 53,000-83,000 by GPC

orbital energy

HOMO -5.22 eV 
LUMO -3.45 eV 

semiconductor properties

P-type (mobility=7×10−4 cm2/V·s)

General description

PBDTTT-CF is a low band-gap conducting polymer which contains dialkoxyl benzodithiophene and fluorine substituted thieno[3,4-b]thiophene groups. It can be used as a donor material with a power conversion efficiency of 6.77%.[1]

Application

Low band gap polymer; for high-efficiency organic solar cells (OPVs) application

OPV Device Structure: ITO/PEDOT:PSS/PBDTTT-CF :PC71BM/Ca/Al; achieving power conversion efficiency as high as 6.77%; as certified by the National Renewable Energy Laboratory (NREL).
PBDTTT-CF is mainly used in the fabrication of polymeric solar cells and organic solar cells.[2][3][4]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and photovoltaic properties of an alternating phenylenevinylene copolymer with substituted-triphenylamine units along the backbone for bulk heterojunction and dye-sensitized solar cells
Mikroyannidis JA, et al.
Journal of Power Sources, 196(4), 2364-2372 (2011)
A polybenzo [1, 2-b: 4, 5-b?] dithiophene derivative with deep HOMO level and its application in high-performance polymer solar cells
Huo L, et al.
Angewandte Chemie (International Edition in English), 122(8), 1542-1545 (2010)
Polymer solar cells with enhanced open-circuit voltage and efficiency
Nature Photonics, 3, 649-653 (2009)
Polymer solar cells with enhanced open-circuit voltage and efficiency
Chen H, et al.
Nature Photonics, 3(11), 649-649 (2009)
Towards organic solar cells without the hole transporting layer on the plasmon-enhanced ITO electrode
Vojtko A, et al.
physica status solidi (a), 212(4), 867-876 (2015)

Articles

The development of high-performance conjugated organic molecules and polymers has received widespread attention in industrial and academic research.

Organic photovoltaics (OPVs) represent a low-cost, lightweight, and scalable alternative to conventional solar cells. While significant progress has been made in the development of conventional bulk heterojunction cells, new approaches are required to achieve the performance and stability necessary to enable commercially successful OPVs.

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service