Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

772267

Sigma-Aldrich

Diethyl 1,2-butadiene-3,4-dicarboxylate

95%

Synonym(s):

2-(Ethenylidene)butanedioic acid 1,4-diethyl ester, Diethyl 2-ethenylidenesuccinate, Diethyl butane-1,2-diene-3,4-dicarboxylate, Ethenylidenebutanedioic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O4
CAS Number:
Molecular Weight:
198.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

liquid

refractive index

n20/D 1.468

density

1.060 g/mL at 25 °C

functional group

ester

storage temp.

2-8°C

SMILES string

CCOC(CC(C(OCC)=O)=C=C)=O

InChI

1S/C10H14O4/c1-4-8(10(12)14-6-3)7-9(11)13-5-2/h1,5-7H2,2-3H3

InChI key

QBOGSIQVGPAAFG-UHFFFAOYSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service