Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

772178

Sigma-Aldrich

Phenyl 2,3,4,6-tetrakis-O-benzyl-1-thio-α-D-mannopyranoside

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H40O5S
CAS Number:
Molecular Weight:
632.81
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

99%

form

solid

refractive index

n20/D 1.596

density

1.142 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[C@@H]1(SC2=CC=CC=C2)[C@@H](OCC3=CC=CC=C3)[C@@H](OCC4=CC=CC=C4)[C@H](OCC5=CC=CC=C5)[C@@H](COCC6=CC=CC=C6)O1

InChI

1S/C40H40O5S/c1-6-16-31(17-7-1)26-41-30-36-37(42-27-32-18-8-2-9-19-32)38(43-28-33-20-10-3-11-21-33)39(44-29-34-22-12-4-13-23-34)40(45-36)46-35-24-14-5-15-25-35/h1-25,36-40H,26-30H2/t36-,37-,38+,39+,40-/m1/s1

InChI key

IKCMSYGNAFDJNX-WUXZAEFSSA-N

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service