Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

772062

Sigma-Aldrich

Fmoc-D-Met-OH

98%

Synonym(s):

Fmoc-D-methionine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21NO4S
CAS Number:
Molecular Weight:
371.45
Beilstein/REAXYS Number:
5384578
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

assay

98%

form

powder or crystals

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

120-124 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

CSCC[C@@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C20H21NO4S/c1-26-11-10-18(19(22)23)21-20(24)25-12-17-15-8-4-2-6-13(15)14-7-3-5-9-16(14)17/h2-9,17-18H,10-12H2,1H3,(H,21,24)(H,22,23)/t18-/m1/s1

InChI key

BUBGAUHBELNDEW-GOSISDBHSA-N

Looking for similar products? Visit Product Comparison Guide

related product

Product No.
Description
Pricing

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Pech et al.
Nucleic acids research, 45(7), 3997-4005 (2017-02-06)
Biological evolution resulted in a homochiral world in which nucleic acids consist exclusively of d-nucleotides and proteins made by ribosomal translation of l-amino acids. From the perspective of synthetic biology, however, particularly anabolic enzymes that could build the mirror-image counterparts

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service