Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

771880

Sigma-Aldrich

Bis(bicyclo[2.2.1]-2-heptyl)chloroborane

Synonym(s):

Bis-norbornylchloroborane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22BCl
CAS Number:
Molecular Weight:
236.59
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

liquid

refractive index

n20/D 1.517

bp

131-136 °C/760 mmHg (decomposition)

density

1.058 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

ClB(C1C[C@@H]2CC[C@H]1C2)C3C[C@@H]4CC[C@H]3C4

InChI

1S/C14H22BCl/c16-15(13-7-9-1-3-11(13)5-9)14-8-10-2-4-12(14)6-10/h9-14H,1-8H2/t9-,10-,11+,12+,13?,14?/m1/s1

InChI key

FIZRDRGODZJEDW-NXAPOWPLSA-N

Application

Reagent promotes diastereoselective syn-aldol products from beta-gamma unsaturated propanoates.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

P Veeraraghavan Ramachandran et al.
Organic letters, 15(6), 1398-1401 (2013-03-08)
It is reported that appropriate dienolborinates can provide access to both diastereomers of 2-(hydroxymethyl)but-3-enoates through exclusive α-regiocontrol in a non-vinylogous pathway. Contrary to previous reports in which dialkylchloroboranes failed to enolize propanoates, acidity-enhanced but-3-enoates readily undergo enolization, offering unprecedented control

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service