Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

771848

Sigma-Aldrich

NVOC2-5-carboxy-Q-rhodamine-NHS ester

95% (HPLC)

Synonym(s):

1′,11′-bis{[(4,5-dimethoxy-2-nitrobenzyl)oxy]carbonyl}-3-oxo-1′,2′,3′,4′,8′,9′,10′,11′-octahydro-spiro[isobenzofuran-1(3H),6′-pyrano[3,2-g:5,6-g′]diquinoline]-5-carboxylic acid, 5-succinimidyl ester, ortho-nitroveratryloxycarbonyl-Q-rhodamine-5-succinimidyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H43N5O19
CAS Number:
Molecular Weight:
1029.91
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Quality Level

assay

95% (HPLC)

form

solid

mp

250-255 °C (decomposition)

storage temp.

−20°C

SMILES string

O=C(C1=C2C=CC(C(ON(C(CC3)=O)C3=O)=O)=C1)OC42C5=C(C=C(N(C(OCC6=CC(OC)=C(OC)C=C6[N+]([O-])=O)=O)CCC7)C7=C5)OC8=C4C=C(CCCN9C(OCC%10=C([N+]([O-])=O)C=C(OC)C(OC)=C%10)=O)C9=C8

InChI

1S/C51H43N5O19/c1-67-41-18-29(37(55(63)64)22-43(41)69-3)24-71-49(61)52-13-5-7-26-16-33-39(20-35(26)52)73-40-21-36-27(8-6-14-53(36)50(62)72-25-30-19-42(68-2)44(70-4)23-38(30)56(65)66)17-34(40)51(33)32-10-9-28(15-31(32)48(60)74-51)47(59)75-54-45(57)11-12-46(54)58/h9-10,15-23H,5-8,11-14,24-25H2,1-4H3

InChI key

ANVQNDABUUZZMB-UHFFFAOYSA-N

Application

Photoactivatable rhodamine derivative functionalized with a succinimidyl ester (NHS). The NHS functionality can be used to conjugate this caged rhodamine to amine containing compounds or biomolecules. The ortho-Nitroveratryloxycarbonyl (NVOC) cages can be removed upon irradiation with UV light, restoring the fluorescence of the base dye. NVOC has an absorption max at ~350 nm and has been shown to be cleaved at 350 nm, 405 nm and 420 nm.

Q-Rhodamine λ(ex) = 537 nm, λ(em) = 556 nm

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K R Gee et al.
Bioorganic & medicinal chemistry letters, 11(16), 2181-2183 (2001-08-22)
An amine-reactive caged rhodamine was synthesized and conjugated to aminodextran. The resulting tracer was injected into a single cell zebrafish embryo, and a portion of the tracer was photolyzed in a single cell after development. The resulting fluorescent cell was
Facile and general synthesis of photoactivatable xanthene dyes.
Laura M Wysocki et al.
Angewandte Chemie (International ed. in English), 50(47), 11206-11209 (2011-09-29)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service