Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

771635

Sigma-Aldrich

2,9-Dipropylanthra[2,1,9-def:6,5,10-def′]diisoquinoline-1,3,8,10(2H,9H)tetrone

97%

Synonym(s):

Bis(n-propylimido)perylene, PDI-C3, Perylene-3,4,9,10-tetracarboxylic acid bis(propylimide)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H22N2O4
CAS Number:
Molecular Weight:
474.51
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

97%

form

solid

mp

>300 °C

λmax

524, 488 nm

semiconductor properties

N-type (mobility=0.1-2.1 cm2/V·s)

SMILES string

CCCN1C(=O)c2ccc3c4ccc5C(=O)N(CCC)C(=O)c6ccc(c7ccc(C1=O)c2c37)c4c56

InChI

1S/C30H22N2O4/c1-3-13-31-27(33)19-9-5-15-17-7-11-21-26-22(30(36)32(14-4-2)29(21)35)12-8-18(24(17)26)16-6-10-20(28(31)34)25(19)23(15)16/h5-12H,3-4,13-14H2,1-2H3

InChI key

GEIYCUVJOKJQPO-UHFFFAOYSA-N

General description

2,9-Dipropylanthra[2,1,9-def:6,5,10-def′]diisoquinoline-1,3,8,10(2H,9H)tetrone belongs to the class of perylenebis(dicarboximide) (PDI) that has an electron transporting character. It can be used as an air-stable n-type semiconductor material in organic field effect transistors. It can also be used as an acceptor molecule in organic photovoltaics.[1][2][3]

Application

High electron transporting character; Perylenebis(dicarboximide)s (PDIs) can be used as air-stable n-type semiconductor materials for organic fieldeffect transistors (OFETs);
Excellent candidates as electron accepting building blocks for organic photovoltaics (OPVs).

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Physical vapor deposited films of a perylene derivative: supramolecular arrangement and thermal stability
Fernandes JD, et al.
Materials Research, 20(4), 882-890 (2017)
Evolution of the Molecular Organization in Bis (n-propylimido) perylene Films under Thermal Annealing
Kam A, et al.
Chemistry of Materials, 10(1), 172-176 (1998)
Role of water in the molecular reorientation on thermal annealing of bis (n-propylimido) perylene films
Kam AP, et al.
Langmuir, 16(3), 1185-1188 (2000)
M Carmen Ruiz Delgado et al.
Journal of the American Chemical Society, 132(10), 3375-3387 (2010-02-20)
Perylene tetracarboxylic diimide (PTCDI) derivatives stand out as one of the most investigated families of air-stable n-type organic semiconductors for organic thin-film transistors. Here, we use density functional theory to illustrate how it is possible to control the charge-transport parameters

Articles

Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.

Fabrication procedure of organic field effect transistor device using a soluble pentacene precursor.

Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service