Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

771562

Sigma-Aldrich

Tetraethylene glycol dimethacrylate

contains ≤1500 ppm MEHQ as inhibitor, 95%

Synonym(s):

PEG bismethacrylate, PEG dimethacrylate, PEG hydrogel

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O[CH2CH2OCH2CH2OCOC(CH3)=CH2]2
CAS Number:
Molecular Weight:
330.37
Beilstein/REAXYS Number:
1803537
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

assay

95%

form

liquid

contains

≤1500 ppm MEHQ as inhibitor

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.465

density

1.082 g/mL at 20 °C (lit.)
1.089 g/mL at 25 °C

storage temp.

−20°C

SMILES string

C=C(C)C(OCCOCCOCCOCCOC(C(C)=C)=O)=O

InChI

1S/C16H26O7/c1-13(2)15(17)22-11-9-20-7-5-19-6-8-21-10-12-23-16(18)14(3)4/h1,3,5-12H2,2,4H3

InChI key

LTHJXDSHSVNJKG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

High purity PEG dimethacrylates are commonly used in PEG-based hydrogels and crosslinking hydrophilic polymers.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mariah S. Hahn
Material Matters, 5(3), 62-62 (2010)
Andrea Kasko
Material Matters, 6(3), 67-67 (2011)
Nívea Regina de Godoy Fróes-Salgado et al.
Odontology, 103(2), 160-168 (2014-04-15)
This study evaluated the effect of the combination of two dimethacrylate-based monomers [bisphenol A diglycidyl dimethacrylate (BisGMA) or bisphenol A ethoxylated dimethacrylate (BisEMA)] with diluents either derived from ethylene glycol dimethacrylate (ethylene glycol dimethacrylate, diethylene glycol dimethacrylate, triethylene glycol dimethacrylate
Jin-Oh You et al.
Biomaterials, 57, 22-32 (2015-05-10)
A principal challenge in wound healing is a lack of cell recruitment, cell infiltration, and vascularization, which occurs in the absence of temporal and spatial cues. We hypothesized that a scaffold that expands due to local changes in pH may
E A Dailing et al.
Journal of dental research, 93(12), 1326-1331 (2014-09-25)
Polymeric dental adhesives require the formation of densely crosslinked network structures to best ensure mechanical strength and durability in clinical service. Monomeric precursors to these materials typically consist of mixtures of hydrophilic and hydrophobic components that potentially undergo phase separation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service