Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

771481

Sigma-Aldrich

N, N′-Dimethyl-3,4,9,10-perylenedicarboximide

98%

Synonym(s):

2,9-Dimethyl-anthra[2,1,9-def:6,5,10-d′e′f′]diisoquinoline-1,3,8,10(2H,9H)-tetrone, MePTC, MePTCDI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H14N2O4
CAS Number:
Molecular Weight:
418.40
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

98%

form

powder

mp

>400 °C

λmax

522 nm in dichloromethane

semiconductor properties

N-type (mobility=10-5 cm2/V·s)

SMILES string

CN1C(=O)c2ccc3c4ccc5C(=O)N(C)C(=O)c6ccc(c7ccc(C1=O)c2c37)c4c56

InChI

1S/C26H14N2O4/c1-27-23(29)15-7-3-11-13-5-9-17-22-18(26(32)28(2)25(17)31)10-6-14(20(13)22)12-4-8-16(24(27)30)21(15)19(11)12/h3-10H,1-2H3

InChI key

PJQYNUFEEZFYIS-UHFFFAOYSA-N

General description

N, N′-Dimethyl-3,4,9,10-perylenedicarboximide (Me-PTCDI) is a perylene derivative that can be used as a thermally stable material in the development of an n-type semiconductor device. It has the capacity to have an ordered growth on a variety of substrates. Its conductivity with respect to the frequency tends to increase at different temperatures.[1][2]

Application

Me-PTCDI can be used in the preparation of the hole transporting layers (HTLs), which can further be utilized in the fabrication of perovskite photovoltaic (PPV) cells.[3][4]
This material is air-stable n-type organic semiconductor. These materials also exhibit high thermal stability. Additionally; PBI derivatives have been used for decades as organic high-performance pigments with color shades from red; via maroon; to black.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

AC electrical conductivity and dielectric relaxation studies on n-type organic thin films of N, N?-Dimethyl-3, 4, 9, 10-perylenedicarboximide (DMPDC)
Qashou SI, et al.
Physica B: Condensed Matter, 525(1), 159-163 (2017)
The photovoltaic effect in a heterojunction of molybdenyl phthalocyanine and perylene dye
Signerski R
Journal of Non-Crystalline Solids, 352(40-41), 4319-4324 (2006)
Hong-Mei Zhao et al.
Journal of the American Chemical Society, 131(43), 15660-15668 (2009-10-29)
Quantum chemical protocols explaining the crystal structures and the visible light absorption properties of 3,4:9,10-perylene tetracarboxylic acid bisimide (PBI) derivates are proposed. Dispersion-corrected density functional theory has provided an intermolecular potential energy of PBI dimers showing several energetically low-lying minima
Interface energetics in organo-metal halide perovskite-based photovoltaic cells
Schulz P, et al.
Energy & Environmental Science, 7(4), 1377-1381 (2014)
The lowest energy Frenkel and charge-transfer excitons in quasi-one-dimensional structures: application to MePTCDI and PTCDA crystals
Hoffmann M, et al.
Chemical Physics, 258(1), 73-96 (2000)

Articles

Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.

Fabrication procedure of organic field effect transistor device using a soluble pentacene precursor.

Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service