771457
Bis[2-(1-isoquinolinyl-N)phenyl-C](2,4-pentanedionato-O2,O4)iridium(III)
98%
Synonym(s):
(piq)2Ir(acac), Bis(1-phenylisoquinoline)(acetylacetonate)iridium(III), Ir(piq)2(acac)
About This Item
Recommended Products
assay
98%
form
powder or crystals
mp
366-370 °C
λmax
341 nm in dichloromethane
476 nm in dichloromethane
fluorescence
λem 315 nm in dichloromethane
SMILES string
CC(=O)[CH-]C(C)=O.[Ir+](c1ccccc1-c2nccc3ccccc23)c4ccccc4-c5nccc6ccccc56
InChI
1S/2C15H10N.C5H7O2.Ir/c2*1-2-7-13(8-3-1)15-14-9-5-4-6-12(14)10-11-16-15;1-4(6)3-5(2)7;/h2*1-7,9-11H;3H,1-2H3;/q;;-1;+1
InChI key
BEACXWMIUQYULI-UHFFFAOYSA-N
Application
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service