Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

771392

Sigma-Aldrich

Bromomethylenemorpholinium bromide

95%

Synonym(s):

4-(Bromomethylene)morpholin-4-ium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9Br2NO
CAS Number:
Molecular Weight:
258.94
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

assay

95%

form

crystals

mp

135-150 °C

SMILES string

[Br-].Br\C=[N+]1\CCOCC1

InChI

1S/C5H9BrNO.BrH/c6-5-7-1-3-8-4-2-7;/h5H,1-4H2;1H/q+1;/p-1

InChI key

KSAUMXGUAZEGEA-UHFFFAOYSA-M

Application

Reagent can be used to convert dimethylthiocarbamates (derived from the corresponding alcohol) into the corresponding bromides.

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Primary Alkyl Bromides from Dimethylthiocarbamates
Moynihan, M.F., Tucker, J.W., Abelt, C.J.
Synthesis, 22, 3565-3568 (2008)
An Improved Synthesis of 6-Deoxyhalo Cyclodextrins via Halomethylenemorpholinium Halides Vilsmeier-Haack Type Reagents
Chmurski, K. and Defaye, J.
Tetrahedron Letters, 38, 7365-7368 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service