771252
4-tert-Butylphenylboronic acid MIDA ester
95%
Synonym(s):
2-[4-(tert-Butyl)phenyl]-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione
About This Item
assay
95%
form
solid
mp
255-260 °C
SMILES string
O=C(CN(C)C1)OB(C2=CC=C(C(C)(C)C)C=C2)OC1=O
InChI
1S/C15H20BNO4/c1-15(2,3)11-5-7-12(8-6-11)16-20-13(18)9-17(4)10-14(19)21-16/h5-8H,9-10H2,1-4H3
InChI key
XERUPHPEYRSGRY-UHFFFAOYSA-N
Application
MIDA Boronates
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service