Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

771244

Sigma-Aldrich

Chloro{2,6-bis[(phenylseleno-Se)methyl]phenyl-C}palladium(II)

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H17ClPdSe2
CAS Number:
Molecular Weight:
557.14
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

impurities

≤1% Dichloromethane

mp

162-221 °C

storage temp.

2-8°C

SMILES string

Cl[Pd]c1c(C[Se]c2ccccc2)cccc1C[Se]c3ccccc3

InChI

1S/C20H17Se2.ClH.Pd/c1-3-10-19(11-4-1)21-15-17-8-7-9-18(14-17)16-22-20-12-5-2-6-13-20;;/h1-13H,15-16H2;1H;/q;;+1/p-1

InChI key

UPTIBUSNQPQCIY-UHFFFAOYSA-M

Application

Application Statement: Complex promotes the formation of alpha-amino acids and allyl alcohols through a Petasis Borono-Mannich reaction.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - STOT RE 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicklas Selander et al.
Journal of the American Chemical Society, 129(44), 13723-13731 (2007-10-13)
An efficient one-pot procedure was designed by integration of the pincer-complex-catalyzed borylation of allyl alcohols in the Petasis borono-Mannich reaction and in allylation of aldehydes and ketones. These procedures are suitable for one-pot synthesis of alpha-amino acids and homoallyl alcohols

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service