771139
(E)-N-(2-Aminoethyl)-4-{2-[4-(3-azidopropoxy)phenyl]diazenyl}benzamide hydrochloride
About This Item
form
solid
reaction suitability
reaction type: click chemistry
reagent type: cross-linking reagent
mp
350-355 °C
Storage temp.
2-8°C
SMILES string
NCCNC(C1=CC=C(C=C1)/N=N/C2=CC=C(OCCCN=[N+]=[N-])C=C2)=O.[H]Cl
InChI
1S/C18H21N7O2.ClH/c19-10-12-21-18(26)14-2-4-15(5-3-14)23-24-16-6-8-17(9-7-16)27-13-1-11-22-25-20;/h2-9H,1,10-13,19H2,(H,21,26);1H/b24-23+;
Inchi Key
UWCLSDSCVCQZQA-XMXXDQCKSA-N
Application
Has been used in proteomic and affinity chromatography applications. The free primary amine can be linked to any carboxylic acid containing compound or biomolecule via standard amide bond forming procedures.The azide can be reacted with a terminal alkyne or cyclooctyne derivative via the 1,3 dipolar cycloaddition click chemistry reaction.
related product
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service