Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

771112

Sigma-Aldrich

(2-Methylpropyl)boronic acid MIDA ester

97%

Synonym(s):

2-Isobutyl-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16BNO4
Molecular Weight:
213.04
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

SMILES string

O=C(CN(C)C1)OB(CC(C)C)OC1=O

InChI

1S/C9H16BNO4/c1-7(2)4-10-14-8(12)5-11(3)6-9(13)15-10/h7H,4-6H2,1-3H3

InChI key

PLRWQFNPZCCLAG-UHFFFAOYSA-N

Application

Protected boronic acid with methylimindodiacetic acid (MIDA) that does not react under standard Suzuki-Miyuara coupling conditions, allowing for selective and iterative cross-coupling sequences. The MIDA boronate also provides prolonged shelf life and stability to typically unstable boronic acid derivatives.

MIDA Boronates

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service