Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

769657

Sigma-Aldrich

Titanium(III,IV) oxide

granular, size 0.8-4 mm

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ti3O5
CAS Number:
Molecular Weight:
223.60
UNSPSC Code:
12352200
NACRES:
NA.23

form

granular

size

0.8-4 mm

impurities

≤0.1% PBB (polybrominated biphenyls)
≤0.1% PBDE (polybrominated diphenyl ethers)

particle size

0.8-4 mm, coeff var >80%

cation traces

Cd: ≤0.01%
Co: ≤0.0005%
Cr: ≤0.005%
Cu: ≤0.001%
Fe: ≤0.02%
Hg: ≤0.1%
Pb: ≤0.1%
V: ≤0.01%

Application

Oxides of titanium find wide range of applications extending from photocatalysis, gas sensing, to antibacterial, protective and anti-reflecting coatings.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Titanium dioxide thin films: preparation and optical properties
Bedikyan , L., et al.
Journal of Chemical Technology and Metallurgy, 48(6), 555-558 (2013)
Fabrication of hydrogen sensors with transparent titanium oxide nanotube-array thin films as sensing elements
Mor, GK., et al.
Thin Solid Films, 496(1), 42-48 (2006)
Titanium dioxide photocatalysis
Fujishima, A., et al.
Journal of Photochemistry and Photobiology, C: Photochemistry Reviews, 1(1), 1-21 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service