Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

769479

Sigma-Aldrich

Ytterbium fluoride

granular, ≤1.6 mm

Synonym(s):

Ytterbium(III) fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
YbF3
CAS Number:
Molecular Weight:
230.04
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.23
form:
granular

form

granular

impurities

≤0.1% O (Oxygen)
≤0.1% PBB (polybrominated biphenyls)
≤0.1% PBDE (polybrominated diphenyl ethers)

particle size

≤1.6 mm
<1.6 mm, coeff var ≥90%

density

~8.20 g/mL at 25 °C (lit.)

cation traces

Cd: ≤0.01%
Co: ≤0.001%
Cr: ≤0.001%
Cu: ≤0.001%
Fe: ≤0.002%
Hg: ≤0.1%
Mn: ≤0.001%
Pb: ≤0.005%

SMILES string

F[Yb](F)F

InChI

1S/3FH.Yb/h3*1H;/q;;;+3/p-3

InChI key

XASAPYQVQBKMIN-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Fluorides of rare earths find applications in anti-reflective coatings. They are also used in synthesis of luminescent bio-imaging probes.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical processing for inorganic fluoride and oxyfluoride materials having optical functions
Fujihara, S. and Tokumo, K.
Journal of Fluorine Chemistry, 130(12), 1106-1110 (2009)
Dual-Modality in Vivo Imaging Using Rare-Earth Nanocrystals With Near-Infrared to Near-Infrared (NIR-to-NIR) Upconversion Luminescence and Magnetic Resonance Properties
Zhou, J., et al.
Biomaterials, 31(12), 3287-3295 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service