Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

76902

Sigma-Aldrich

1-(3-Cyanopropyl)-3-methylimidazolium dicyanamide

≥98.5% (HPLC/T)

Synonym(s):

1-Methyl-3-(3-cyanopropyl)imidazolium dicyanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N6
CAS Number:
Molecular Weight:
216.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥98.5% (HPLC/T)

form

liquid

impurities

≤0.5% water

refractive index

n20/D 1.524

SMILES string

N#C[N-]C#N.C[n+]1ccn(CCCC#N)c1

InChI

1S/C8H12N3.C2N3/c1-10-6-7-11(8-10)5-3-2-4-9;3-1-5-2-4/h6-8H,2-3,5H2,1H3;/q+1;-1

InChI key

GPLYXNDEHINOHN-UHFFFAOYSA-N

Application

Nitrile-functionalized ionic liquid showing superior characteristics in Suzuki and Stille coupling reactions[1].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongbin Zhao et al.
Journal of the American Chemical Society, 126(48), 15876-15882 (2004-12-02)
A series of relatively low-cost ionic liquids, based on the N-butyronitrile pyridinium cation [C(3)CNpy](+), designed to improve catalyst retention, have been prepared and evaluated in Suzuki and Stille coupling reactions. Depending on the nature of the anion, these salts react

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service