768561
Di-μ-chlorotetrakis[2-(1-isoquinolinyl-N)phenyl-C]diiridium(III)
95% (HPLC)
Synonym(s):
Iridium phenylquinoline dichloro-bridged dimer, [(pq)2IrCl]2
About This Item
Recommended Products
assay
95% (HPLC)
form
solid
mp
440-455 °C
SMILES string
Cl[Ir].Cl[Ir].c1ccc(cc1)-c2nccc3ccccc23.c4ccc(cc4)-c5nccc6ccccc56.c7ccc(cc7)-c8nccc9ccccc89.c%10ccc(cc%10)-c%11nccc%12ccccc%11%12
InChI
1S/4C15H11N.2ClH.2Ir/c4*1-2-7-13(8-3-1)15-14-9-5-4-6-12(14)10-11-16-15;;;;/h4*1-11H;2*1H;;/q;;;;;;2*+1/p-2
InChI key
ZUKCRERFHSESIM-UHFFFAOYSA-L
General description
The applications of such Ir complexes are based on the fact that neutral Ir based complexes consist of distinct carrier transporting/blocking and emitting layers. Single active layer in cationic Ir complexes is responsible for both carrier transport and light emission. Studies on anionic Ir are mainly focused on their photophysics.[1][2]
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service