Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

768383

Sigma-Aldrich

Zirconium(IV) oxide-yttria stabilized

greener alternative

nanopowder, 99.99% trace metals basis, contains 40 mol % yttria as stabilizer

Synonym(s):

40YSZ, YSZ40, YSZ, Yttria stabilized zirconia, Yttrium stabilized zirconium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
(ZrO2)0.60(Y2O3)0.40
UNSPSC Code:
12352200

assay

99.99% trace metals basis

form

nanopowder

contains

40 mol % yttria as stabilizer

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

greener alternative category

General description

Yttria stabilized zirconia (YSZ) is the most commonly used solid electrolyte for solid oxide fuel cells (SOFCs) and oxygen sensors.[1] YSZ exhibits high oxygen-ionic conductivity while blocking electronic conduction.[2] YSZ particles may be synthesized from yttrium nitrate or isopropoxide by sol gel method.[3] YSZ nanoparticles may be prepared in supercritical water (SCW).[4] YSZ nanoparticles of different sizes with varying % mol of yttria may be obtained by CO2 laser based continuous/pulsed laser vaporization method.[5]
Zirconium(IV) oxide-yttria stabilized (YSZ40) (nanopowder, 99.99% trace metals basis, contains 40 mol % yttria as stabilizer) is an oxygen ion conductor with properties such as good ionic conductivity and high physiochemical stability. Its other properties include excellent shock resistance, low thermal conductivity and a high coefficient of thermal expansion.[6][7][8]

Application

Collagen-yttria-stabilized amorphous zirconia hybrid scaffolds may be developed.[2] YSZ thin films may be used as thermal barrier coatings on combustors, stationary and rotating components of gas turbines or diesel engines. 6
YSZ can be used for a variety of applications such as:
  • a thermal barrier coating for the protection and insulation of metal components in gas turbines[7]
  • an electrolyte in the fabrication of solid oxide fuel cells (SOFCs)[9]
  • in the fabrication of zirconia dental implant abutments[10]
  • membranes for micro energy conversion devices[11]
  • a solid acid and a catalyst for catalytic applications[12]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Goodenough, J. B.
Annual Review of Materials Research, 33, 91-91 (2003)
Shim; J. H.;
Acta Materialia, 60(1), 1-1 (2012)
De Souza; S.;
Journal of the Electrochemical Society, 144(3), L35-L35 (1997)
Fracture resistance of yttria-stabilized zirconia dental implant abutments
Adatia ND, et al.
Journal of Prosthodontics : Official Journal of the American College of Prosthodontists, 18(1), 17-22 (2009)
Confined Transformation of UiO-66 Nanocrystals to Yttria-Stabilized Zirconia with Hierarchical Pore Structures for Catalytic Applications
Qin R and Zeng H
Advances in Functional Materials, 29(39), 1903264-1903264 (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service