Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

768235

Sigma-Aldrich

Dibromobis(acetonitrile)palladium(II)

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6Br2N2Pd
CAS Number:
Molecular Weight:
348.33
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

95%

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

214-263 °C (decomposition)

SMILES string

CC#N.CC#N.Br[Pd]Br

InChI

1S/2C2H3N.2BrH.Pd/c2*1-2-3;;;/h2*1H3;2*1H;/q;;;;+2/p-2

InChI key

CLLBUWJGRMZLBR-UHFFFAOYSA-L

Application

  • Versatile palladium precursor

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service