768065
2-(Diphenylphosphino)-N,N-dimethylbenzylamine
95%
Synonym(s):
(2-((Dimethylamino)methyl)phenyl)diphenylphosphine
About This Item
Recommended Products
assay
95%
form
liquid
reaction suitability
reagent type: ligand
reaction type: Cross Couplings
mp
55-59 °C
functional group
phosphine
SMILES string
CN(C)CC1=C(C=CC=C1)P(C2=CC=CC=C2)C3=CC=CC=C3
InChI
1S/C21H22NP/c1-22(2)17-18-11-9-10-16-21(18)23(19-12-5-3-6-13-19)20-14-7-4-8-15-20/h3-16H,17H2,1-2H3
InChI key
POILNYUNIDNVDD-UHFFFAOYSA-N
Application
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service