Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

767573

Sigma-Aldrich

Poly(ethylene glycol) bis(2-bromoisobutyrate)

average Mn 700, cross-linking reagent polymerization reactions, bromo

Synonym(s):

Polyethylene glycol, Dibromoisobutyryl PEG, PEG-ATRP, PEG-Bib

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

Product Name

Poly(ethylene glycol) bis(2-bromoisobutyrate), average Mn 700

form

liquid

mol wt

PEG average Mn 300 (n ~ 7)
average Mn 700

reaction suitability

reagent type: cross-linking reagent
reaction type: Polymerization Reactions

Ω-end

bromo

α-end

bromo

polymer architecture

shape: linear
functionality: homobifunctional

storage temp.

2-8°C

Application

PEG functionalized with bromoisobutyryl group to initiate atom transfer radical polymerization (ATRP). This macroinitiator can be used to increase biocompatibility; hydrophilicity; or generate a triblock copolymer with PEG as the middle block.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service