767468
2,9-Bis[2-(4-chlorophenyl)ethyl]anthra[2,1,9-def:6,5,10-d′e′f′]diisoquinoline-1,3,8,10(2H,9H)tetrone
98%
Synonym(s):
4ClPEPTC, N,N′-Bis[2-(4-chlorophenyl)ethyl]-3,4,9,10-perylenetetradicarboximide
About This Item
assay
98%
form
solid
mp
393-397 °C
SMILES string
Clc1ccc(CCN2C(=O)c3ccc4c5ccc6C(=O)N(CCc7ccc(Cl)cc7)C(=O)c8ccc(c9ccc(C2=O)c3c49)c5c68)cc1
InChI
1S/C40H24Cl2N2O4/c41-23-5-1-21(2-6-23)17-19-43-37(45)29-13-9-25-27-11-15-31-36-32(40(48)44(39(31)47)20-18-22-3-7-24(42)8-4-22)16-12-28(34(27)36)26-10-14-30(38(43)46)35(29)33(25)26/h1-16H,17-20H2
InChI key
QSDATNMVYLQQKT-UHFFFAOYSA-N
Application
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Fabrication procedure of organic field effect transistor device using a soluble pentacene precursor.
Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.
Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service