Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

767379

Sigma-Aldrich

Poly(acrylamide)

average Mn 150,000

Synonym(s):

Polyacrylamide, PAAm, PAM, Poly(2-Propenamide)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C3H5NO)n
CAS Number:
MDL number:
UNSPSC Code:
12352200

form

solid

mol wt

Mn 150,000 (typical)
Mw 400,000 (typical)
Mw/Mn 2-3
average Mn 150,000

composition

EtOH, ≤20%

impurities

≤20% EtOH (residual solvent)

mp

252-257 °C

Mw/Mn

2‑3

storage temp.

2-8°C

InChI

1S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)

InChI key

HRPVXLWXLXDGHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Polyacrylamide is commonly used as a water soluble homopolymer which is biocompatible.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sachio Yamamoto
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 132(9), 1031-1035 (2012-10-02)
Microchip electrophoresis is widely used for microfluidics and has been studied extensively over the past decade. Translation of capillary electrophoresis methods from traditional capillary systems to a microchip platform provides rapid separation and easy quantitation of sample components. However, most
D J King et al.
Reviews on environmental health, 8(1-4), 3-16 (1989-01-01)
The purpose of this paper is to present information gathered regarding, in general, the physical characteristics, and, in particular, the possible toxic nature of polyacrylamide A short discussion of the properties and toxicity of the acrylamide monomer is also included.
Imran Khimji et al.
Methods (San Diego, Calif.), 64(3), 292-298 (2013-08-28)
Biosensors are devices that can provide quantitative or semi-quantitative analytical information about target molecules, where molecular recognition is based on biomolecular interactions. In recent years, DNA has emerged as a useful molecule for biosensor development since DNA can not only
N V Bovin
Bioorganicheskaia khimiia, 22(9), 643-663 (1996-09-01)
The synthesis, physicochemical characteristics, and application for studying the carbohydrate-binding molecules of some analogues of human cell glycoconjugates, neoglycoconjugates, were reviewed. An approach to the synthesis of the polyacrylamide derivatives of carbohydrates based on the interaction of fully activated polyacrylic
E A Smith et al.
Reviews on environmental health, 9(4), 215-228 (1991-10-01)
Acrylamide is a highly water soluble vinyl monomer formed from the hydration of acrylonitrile. The major commercial use of acrylamide is the formation of polymers. In the environment acrylamide has a high mobility in soil, may travel great distances in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service