Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

767328

Sigma-Aldrich

Polyester bis-MPA dendron, 2 hydroxyl, 1 acetylene

generation 1, 95%

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

description

hydroxyl surface groups

assay

95%

form

chunks

mol wt

generation 1

feature

focal point acetylene

no. Surface Groups

2

mp

45-50 °C

Storage temp.

2-8°C

SMILES string

CC(CO)(CO)C(=O)OCC#C

InChI

1S/C8H12O4/c1-3-4-12-7(11)8(2,5-9)6-10/h1,9-10H,4-6H2,2H3

Inchi Key

RJGDJCXFZLEVMD-UHFFFAOYSA-N

General description

calculated mol. wt. 172.2

Application

Acetylene dendrons can be coupled to azide functional molecules such as carbohydrates, fluorescent dyes and polymers.

Other Notes

Bis-MPA = 2,2-Bis(hydroxyl-methyl)propionic acid

Legal Information

Manufactured by Polymer Factory Sweden AB

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service