767212
Polyester bis-MPA dendron, 4 hydroxyl, 1 azide
generation 2, 95%
Synonym(s):
bisMPA dendrimer, bisMPA dendron
About This Item
description
hydroxyl surface groups
assay
95%
form
solid
mol wt
generation 2
feature
focal point azide
no. Surface Groups
4
mp
93-98 °C (decomposition)
storage temp.
−20°C
SMILES string
CC(CO)(CO)C(=O)OCC(C)(COC(=O)C(C)(CO)CO)C(=O)OCCCCCCN=[N+]=[N-]
InChI
1S/C21H37N3O10/c1-19(10-25,11-26)16(29)33-14-21(3,15-34-17(30)20(2,12-27)13-28)18(31)32-9-7-5-4-6-8-23-24-22/h25-28H,4-15H2,1-3H3
InChI key
LGPUUVBKCMNFLE-UHFFFAOYSA-N
General description
application
- coatings
- resins
- additives,
- analytical techniques
- medicine
- drug and dye delivery
- molecular imprinting
- organic polymer synthesis
- catalysis
- optoelectronic applications.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service