Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

767158

Sigma-Aldrich

Polyester bis-MPA dendron, 32 carboxyl, 1 amine

generation 5

Synonym(s):

bisMPA dendrimer, bisMPA dendron

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
PubChem Substance ID:

description

carboxyl surface groups

form

solid

mol wt

generation 5

feature

focal point amine

no. Surface Groups

32

mp

>300 °C

Storage temp.

−20°C

Inchi Key

NQULMOAGCWOIQE-UHFFFAOYSA-N

Application

Amine dendrons can be covalently attached through conjugation to carboxyl groups, primary amines, or sulfhydryl groups. Amines can also attach to oxidized carbohydrate groups (usually aldehydes) by reductive amination.

Other Notes

InChI: 1S/C288H385NO186.C4H6O4/c1-33-258(3,101-414-196(352)70-39-165(290)291)228(384)446-132-274(19,133-447-229(385)259(4,102-415-197(353)71-40-166(292)293)103-416-198(354)72-41-167(294)295)244(400)462-148-282(27,149-463-245(401)275(20,134-448-230(386)260(5,104-417-199(355)73-42-168(296)297)105-418-200(356)74-43-169(298)299)135-449-231(387)261(6,106-419-201(357)75-44-170(300)301)107-420-202(358)76-45-171(302)303)252(408)470-156-286(31,157-471-253(409)283(28,150-464-246(402)276(21,136-450-232(388)262(7,108-421-203(359)77-46-172(304)305)109-422-204(360)78-47-173(306)307)137-451-233(389)263(8,110-423-205(361)79-48-174(308)309)111-424-206(362)80-49-175(310)311)151-465-247(403)277(22,138-452-234(390)264(9,112-425-207(363)81-50-176(312)313)113-426-208(364)82-51-177(314)315)139-453-235(391)265(10,114-427-209(365)83-52-178(316)317)115-428-210(366)84-53-179(318)319)256(412)474-161-288(34-2,160-445-227(383)163-35-37-164(289)38-36-163)162-475-257(413)287(32,158-472-254(410)284(29,152-466-248(404)278(23,140-454-236(392)266(11,116-429-211(367)85-54-180(320)321)117-430-212(368)86-55-181(322)323)141-455-237(393)267(12,118-431-213(369)87-56-182(324)325)119-432-214(370)88-57-183(326)327)153-467-249(405)279(24,142-456-238(394)268(13,120-433-215(371)89-58-184(328)329)121-434-216(372)90-59-185(330)331)143-457-239(395)269(14,122-435-217(373)91-60-186(332)333)123-436-218(374)92-61-187(334)335)159-473-255(411)285(30,154-468-250(406)280(25,144-458-240(396)270(15,124-437-219(375)93-62-188(336)337)125-438-220(376)94-63-189(338)339)145-459-241(397)271(16,126-439-221(377)95-64-190(340)341)127-440-222(378)96-65-191(342)343)155-469-251(407)281(26,146-460-242(398)272(17,128-441-223(379)97-66-192(344)345)129-442-224(380)98-67-193(346)347)147-461-243(399)273(18,130-443-225(381)99-68-194(348)349)131-444-226(382)100-69-195(350)351;5-3(6)1-2-4(7)8/h35-38H,33-34,39-162,289H2,1-32H3,(H,290,291)(H,292,293)(H,294,295)(H,296,297)(H,298,299)(H,300,301)(H,302,303)(H,304,305)(H,306,307)(H,308,309)(H,310,311)(H,312,313)(H,314,315)(H,316,317)(H,318,319)(H,320,321)(H,322,323)(H,324,325)(H,326,327)(H,328,329)(H,330,331)(H,332,333)(H,334,335)(H,336,337)(H,338,339)(H,340,341)(H,342,343)(H,344,345)(H,346,347)(H,348,349)(H,350,351);1-2H2,(H,5,6)(H,7,8)
Bis-MPA = 2,2-Bis(hydroxyl-methyl)propionic acid

Legal Information

Manufactured by Polymer Factory Sweden AB

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service