Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

767115

Sigma-Aldrich

Polyester bis-MPA dendron, 32 hydroxyl, 1 thiol

generation 5, 95%

Synonym(s):

bisMPA dendrimer, bisMPA dendron

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002

description

hydroxyl surface groups

assay

95%

form

solid

mol wt

generation 5

feature

focal point thiol

no. Surface Groups

32

mp

>300 °C

storage temp.

−20°C

General description

calculated mol. wt. 3677.7

Application

Thiol dendrons can be conjugated to varius noble surfaces or coupled to unsaturated carbon-carbon bonds using various radical initiators.

Other Notes

Bis-MPA = 2,2-Bis(hydroxyl-methyl)propionic acid

Legal Information

Manufactured by Polymer Factory Sweden AB

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service