Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

766763

Sigma-Aldrich

1-(Z-Amino)-1-(aminomethyl)cyclopentane hydrochloride

97%

Synonym(s):

1-(Cbz-Amino)-1-(aminomethyl)cyclopentane hydrochloride, Benzyl [1-(aminomethyl)cyclopentyl]carbamate hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20N2O2 · HCl
CAS Number:
Molecular Weight:
284.78
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

178-181 °C

functional group

amine
phenyl

SMILES string

NCC1(NC(OCC2=CC=CC=C2)=O)CCCC1.Cl

InChI

1S/C14H20N2O2.ClH/c15-11-14(8-4-5-9-14)16-13(17)18-10-12-6-2-1-3-7-12;/h1-3,6-7H,4-5,8-11,15H2,(H,16,17);1H

InChI key

JHHQXOFHNQWXMN-UHFFFAOYSA-N

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service