Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

766631

Sigma-Aldrich

mono-Iodoborane triethylamine complex solution

0.6 M in dichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H17BIN
Molecular Weight:
240.92
UNSPSC Code:
12352116
PubChem Substance ID:

form

liquid

concentration

0.6 M in dichloromethane

density

1.341 g/mL at 25 °C

functional group

amine

storage temp.

2-8°C

SMILES string

BI.CCN(CC)CC

InChI

1S/C6H15N.BH2I/c1-4-7(5-2)6-3;1-2/h4-6H2,1-3H3;1H2

InChI key

DMHTWFRWCXBOCV-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

122.0 °F

flash_point_c

50 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service