Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

766585

Sigma-Aldrich

1,3-Dibromo-5-octyl-4H-thieno[3,4-c]pyrrole-4,6(5H)-dione

≥99.5% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17Br2NO2S
CAS Number:
Molecular Weight:
423.16
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

≥99.5% (HPLC)

form

solid

mp

105-109 °C

SMILES string

CCCCCCCCN1C(=O)c2c(Br)sc(Br)c2C1=O

InChI

1S/C14H17Br2NO2S/c1-2-3-4-5-6-7-8-17-13(18)9-10(14(17)19)12(16)20-11(9)15/h2-8H2,1H3

InChI key

GSGMEQUXTCYOAU-UHFFFAOYSA-N

General description

1,3-Dibromo-5-octyl-4H-thieno[3,4-c]pyrrole-4,6(5H)-dione is a thienopyrrolodione (TPD) based electron acceptor material (n-type semiconductor) which is used widely for organic photovoltaic (OPV) applications. They have very powerful electron withdrawing capability. TPD based conjugated polymers have exhibited a Power Conversion Efficiency (PCE) of as high as 7.3% in bulk heterojunction polymer solar cells.[1]

Application

Used as an electron acceptor material (n-type semiconductor) in Polymer Solar Cells.[2]

Features and Benefits

High electron withdrawing capability, deeper HUMO levels leading to high Voc.[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bridget Carsten et al.
Journal of the American Chemical Society, 133(50), 20468-20475 (2011-11-15)
A new low band gap copolymer PBB3 containing [6,6']bi[thieno[3,4-b]thiophenyl]-2,2'-dicarboxylic acid bis-(2-butyloctyl) ester (BTT) and 4,8-bis(2-butyloctyl)benzo[1,2-b:4,5-b']dithiophene (BDT) units was synthesized and tested for solar cell efficiency. PBB3 showed a broad absorbance in the near-IR region with a substantially red-shifted (by more
Conjugated polymers based on 1, 3-dithien-2-yl-thieno [3, 4-c] pyrrole-4, 6-dione: synthesis, characterization, and solvent effects on photovoltaic performance
Zhou, Erjun, et al.
The Journal of Physical Chemistry C, 116(3), 2608-2614 (2012)
Alternating Copolymers Consisting of Dialkoxylated Naphthalene along with Either Dithiophenylated 1, 4-Dioxopyrrolo-[3, 4c]-pyrrole or 5H-thieno [3, 4-c] pyrrole-4, 6-dione: Synthesis and Photo-Electrochemical Properties
Lim, Young Lae, et al.
Bull. Korean Chem. Soc., 34(1), 313-316 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service