Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

766569

Sigma-Aldrich

Poly(sulfopropyl acrylate) potassium salt

average Mn 200,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(C6H9O2SO3K)nH
UNSPSC Code:
12162002
NACRES:
NA.23

mol wt

average Mn 200,000

viscosity

60 cP, 1 % in H2O(25 °C)

transition temp

Tm 358-363 °C

Mw/Mn

~2

storage temp.

2-8°C

General description

The three dimensional flexible network can absorb and retain aqueous solution
without dissolving thus forming superhydrogels.

Application

It may find applications in:
1.controlled drug release systems
2.thermoresponsive hydrogels

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service