Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

766399

Sigma-Aldrich

2-Bromoisobutyric anhydride

95%

Synonym(s):

2-Bromo-2-methylpropanoic anhydride, ATRP initiator

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12Br2O3
CAS Number:
Molecular Weight:
315.99
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

assay

95%

form

powder

mp

56-60 °C

SMILES string

CC(C)(Br)C(=O)OC(=O)C(C)(C)Br

InChI

1S/C8H12Br2O3/c1-7(2,9)5(11)13-6(12)8(3,4)10/h1-4H3

InChI key

CHYAZMNKLHWVBP-UHFFFAOYSA-N

Application

Highly effective difunctional initiator for atom transfer radical polymerization (ATRP).

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Xiujun Ren et al.
Journal of chromatography. A, 1618, 460904-460904 (2020-01-30)
A novel chiral stationary phase (CSP) was prepared through the reaction of surface-initiated atom transfer radical polymerization (ATRP) by the copolymerization of thermoresponsive N-isopropylacrylamide (NIPAM) and β-cyclodextrin (β-CD) on the silica beads for high performance liquid chromatography (HPLC). X-ray photoelectron
Shuaidong Huo et al.
Nature chemistry, 13(2), 131-139 (2021-01-31)
Pharmaceutical drug therapy is often hindered by issues caused by poor drug selectivity, including unwanted side effects and drug resistance. Spatial and temporal control over drug activation in response to stimuli is a promising strategy to attenuate and circumvent these

Articles

Over the past two decades, the rapid advance of controlled living polymerization (CLP) techniques.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service