Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

766208

Sigma-Aldrich

Diphenylammonium Triflate

greener alternative

97%

Synonym(s):

DPAT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12F3NO3S
CAS Number:
Molecular Weight:
319.30
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

reaction suitability

reagent type: ligand

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

178-183 °C

greener alternative category

storage temp.

2-8°C

SMILES string

[O-]S(=O)(=O)C(F)(F)F.[NH2+](c1ccccc1)c2ccccc2

InChI

1S/C12H11N.CHF3O3S/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;2-1(3,4)8(5,6)7/h1-10,13H;(H,5,6,7)

InChI key

MGEGQAUINMTPGX-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Efficient catalyst for esterification of carboxylic acids and for transesterification of carboxylic
esters with alcohols
Efficient catalyst for greener synthesis of esters by condensation of carboxylic acids with alcohols or transfesterification.
Diphenylammonium triflate (DPAT): efficient catalyst for esterification of carboxylic acids and for transesterification of carboxylic esters with nearly equimolar amounts of alcohols

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

José-Ángel García-Pedraza et al.
Clinical and experimental pharmacology & physiology, 44(12), 1224-1231 (2017-08-05)
5-hydroxytryptamine (5-HT) modulates noradrenergic activity in different cardiovascular territories, but its effect on the mesenteric vasopressor outflow has not yet been clarified. This study investigated the in vivo serotonergic influence, characterizing 5-HT receptors implicated, in sympathetic innervation of mesenteric vasculature.
Julian Maier et al.
Neuropharmacology, 138, 282-291 (2018-06-17)
(±)-cis-4,4'-Dimethylaminorex (4,4'-DMAR) is a new psychoactive substance (NPS) that has been associated with 31 fatalities and other adverse events in Europe between June 2013 and February 2014. We used in vitro uptake inhibition and transporter release assays to determine the effects
Sarah Delcourte et al.
CNS neuroscience & therapeutics, 23(6), 518-525 (2017-04-19)
Asenapine is a new atypical antipsychotic prescribed for the treatment of psychosis/bipolar disorders that presents higher affinity for serotonergic than dopaminergic receptors. The objective of this study was to investigate its antidepressant-like and antimanic-like properties on relevant animal models of
Tetrahedron Letters, 41, 5249-5252 (2000)
T M Eriksson et al.
Molecular psychiatry, 17(2), 173-184 (2011-01-19)
Cognitive dysfunctions are common in major depressive disorder, but have been difficult to recapitulate in animal models. This study shows that Flinders sensitive line (FSL) rats, a genetic rat model of depression, display a pronounced impairment of emotional memory function

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service