Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

766089

Sigma-Aldrich

Phenylselenozinc chloride

Synonym(s):

(Phenylselanyl)zinc(II) chloride, Benzeneselenolato zinc cloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5ClSeZn
CAS Number:
Molecular Weight:
256.91
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

315-325 °C

storage temp.

2-8°C

SMILES string

Cl[Zn][Se]c1ccccc1

InChI

1S/C6H6Se.ClH.Zn/c7-6-4-2-1-3-5-6;;/h1-5,7H;1H;/q;;+2/p-2

InChI key

DICKYPLTHDZRSN-UHFFFAOYSA-L

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Ox. Sol. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service