764833
Poly(lactide-co-caprolactone)-block-poly(ethylene glycol)-block-poly(lactide-co-caprolactone)
average Mn 5700-4600-5700
Synonym(s):
PLCL-PEG-PLCL
About This Item
Recommended Products
form
pellets
feed ratio
lactide 50:50
mol wt
PEG average Mn 5,000 (DCM, PEO)
PLCL average Mn 5,700
average Mn (1,000-10,000-1,000)
degradation timeframe
1-2 months
transition temp
Tm 34-41 °C
PDI
<1.2
<1.3 (typical PEG; overall PDI <1.8 (THF, PEO))
storage temp.
2-8°C
General description
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Local delivery of bioactive molecules using an implantable device can decrease the amount of drug dose required as well as non-target site toxicities compared to oral or systemic drug administration.
AliAliphatic polyesters, including polylactide and polyglycolide, are biodegradable polymers widely used in medical applications.
Humankind has utilized protein materials throughout its existence, starting with the use of materials such as wool and silk for warmth and protection from the elements and continuing with the use of recombinant DNA techniques to synthesize proteins with unique and useful properties.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service