Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

764450

Sigma-Aldrich

α-(Fluoromethyl)benzylamine

95%

Synonym(s):

α-(Fluoromethyl)benzenemethanamine, 2-Fluoro-1-phenylethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10FN
CAS Number:
Molecular Weight:
139.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

liquid

refractive index

n20/D 1.519

density

1.096 g/mL at 25 °C

functional group

amine
fluoro
phenyl

SMILES string

NC(CF)c1ccccc1

InChI

1S/C8H10FN/c9-6-8(10)7-4-2-1-3-5-7/h1-5,8H,6,10H2

InChI key

KSWGCLOZHMSHGB-UHFFFAOYSA-N

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

192.0 °F - closed cup

flash_point_c

88.9 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service