Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

764442

Sigma-Aldrich

4-Bromo-α-(fluoromethyl)benzylamine

97%

Synonym(s):

1-(4-Bromophenyl)-2-fluoroethanamine, 4-Bromo-α-(fluoromethyl)benzenemethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9BrFN
CAS Number:
Molecular Weight:
218.07
UNSPSC Code:
12352100
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.560

mp

214-230 °C

density

1.494 g/mL at 25 °C

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service