Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

764353

Supelco

Trimethyl phosphate solution

25% in chloroform-d (99.8 atom % D), TMS 1 %, NMR tube size 5 mm × 7 in.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12142201
NACRES:
NA.24

composition

TMS, 1%

concentration

25% in chloroform-d (99.8 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 7 in.

suitability

suitable for NMR (reference standard)

format

single component solution

Features and Benefits

31P sensitivity

Quantity

5 mm O.D. tube contains 0.700 mL.

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

target_organs

Central nervous system, Liver,Kidney

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service