Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

764035

Sigma-Aldrich

2-Bromo-5-dodecylthiophene

98% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H27BrS
CAS Number:
Molecular Weight:
331.35
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

98% (GC)

form

liquid

refractive index

n20/D 1.510

density

1.105 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCc1ccc(Br)s1

InChI

1S/C16H27BrS/c1-2-3-4-5-6-7-8-9-10-11-12-15-13-14-16(17)18-15/h13-14H,2-12H2,1H3

InChI key

MOSAVYMGWTYDNS-UHFFFAOYSA-N

Application

Terminal Building Blocks for semiconducting polymers and oligomers to increase solubility and mobility

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic semiconductors for field-effect transistors (FETs): tuning of spectroscopic, electrochemical, electronic and structural properties of naphthalene bisimides via substituents containing alkylthienyl moieties
Journal of Materials Chemistry, 20, 1913-1920 (2010)
Structural-Property Relationship in Pyrazino[2,3-g]quinoxaline Derivatives: Morphology, Photophysical, and Waveguide Properties
Chemistry of Materials, 22, 3735-3745 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service