763942
2,9-Bis[2-(4-fluorophenyl)ethyl]anthra[2,1,9-def:6,5,10-d′e′f′]diisoquinoline-1,3,8,10(2H,9H)tetrone
95%
Synonym(s):
4FPEPTC, N,N′-Bis[2-(4-fluoro-phenyl)-ethyl]- 3,4,9,10-perylenetetradicarboximide
About This Item
assay
95%
form
solid
mp
371-376 °C
SMILES string
Fc1ccc(CCN2C(=O)c3ccc4c5ccc6C(=O)N(CCc7ccc(F)cc7)C(=O)c8ccc(c9ccc(C2=O)c3c49)c5c68)cc1
InChI
1S/C40H24F2N2O4/c41-23-5-1-21(2-6-23)17-19-43-37(45)29-13-9-25-27-11-15-31-36-32(40(48)44(39(31)47)20-18-22-3-7-24(42)8-4-22)16-12-28(34(27)36)26-10-14-30(38(43)46)35(29)33(25)26/h1-16H,17-20H2
InChI key
NTVKTKREOVPUHX-UHFFFAOYSA-N
General description
(a) their structures are well defined and exhibit no molecular weight dependence, leading to improved purity and limiting variation; and
(b) they exhibit more organized nanostructures, leading to higher charge carrier mobility.
Additionally, small molecule architectures are sensitive to minute structure changes; thus, properties like electronic energy levels, optical absorption, and self-assembly tendencies can be systemically tuned to maximize device performance.[1]
Application
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Fabrication procedure of organic field effect transistor device using a soluble pentacene precursor.
Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.
Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service