Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

763667

Sigma-Aldrich

(2,4-Pentanedionato-O2,O4)bis[2-(1H -pyrazol-1-yl-N2)phenyl-C]iridium(III)

96%

Synonym(s):

(PPZ)2Ir(acac)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H21IrN4O2
CAS Number:
Molecular Weight:
577.66
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

96%

form

solid

mp

309-314 °C

λmax

244 nm in chloroform

fluorescence

λex 286 nm; λem 317 nm in THF

SMILES string

CC(=O)\C=C(\C)O[Ir](c1ccccc1-n2cccn2)c3ccccc3-n4cccn4

InChI

1S/2C9H7N2.C5H8O2.Ir/c2*1-2-5-9(6-3-1)11-8-4-7-10-11;1-4(6)3-5(2)7;/h2*1-5,7-8H;3,6H,1-2H3;/q;;;+1/p-1/b;;4-3-;

InChI key

GQRGPQFRJLRFRL-DVACKJPTSA-M

Application

(PPZ)2Ir(acac) can be used in the fabrication of green electrophosphorescent devices. Shows efficiency of up to 9.5 cd/A.[1]

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wu; Li-Lan; et al.
Journal of Luminescence, 126, 687-694 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service