Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

763659

Sigma-Aldrich

3-(Allyloxy)-1-propyne

97%

Synonym(s):

3-(2-Propyn-1-yloxy)-1-propene, 3-(2-Propynyloxy)-1-propene, 3-Propargyloxy-1-propene, Allyl 2-propynyl ether, Allyl propargyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O
Molecular Weight:
96.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

form

powder

refractive index

n20/D 1.431

density

0.8621 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C#CCOCC=C

InChI

1S/C6H8O/c1-3-5-7-6-4-2/h1,4H,2,5-6H2

InChI key

GAQOILKTBJWUQM-UHFFFAOYSA-N

pictograms

FlameSkull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

51.8 °F

flash_point_c

11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service