Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

763586

Sigma-Aldrich

N,N,N′,N′-Tetrakis(2-naphthyl)benzidine

≥98% (HPLC)

Synonym(s):

Β-TNB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H36N2
CAS Number:
Molecular Weight:
688.86
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

≥98% (HPLC)

form

powder or crystals

mp

223-228 °C

λmax

355 nm in THF

SMILES string

c1ccc2cc(ccc2c1)N(c3ccc(cc3)-c4ccc(cc4)N(c5ccc6ccccc6c5)c7ccc8ccccc8c7)c9ccc%10ccccc%10c9

InChI

1S/C52H36N2/c1-5-13-43-33-49(29-21-37(43)9-1)53(50-30-22-38-10-2-6-14-44(38)34-50)47-25-17-41(18-26-47)42-19-27-48(28-20-42)54(51-31-23-39-11-3-7-15-45(39)35-51)52-32-24-40-12-4-8-16-46(40)36-52/h1-36H

InChI key

QKCGXXHCELUCKW-UHFFFAOYSA-N

Application

This material is used as a high performance host material for OLED devices showing increased efficiency (external electroluminescence efficiency of ~12%).[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ayataka Endo and Chihaya Adachi.
Jpn J. App. Phys., Part I, 49, 050205-050205 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service